| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013054.hg.1 |
ENST00000615411.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013054.hg.1 |
ENST00000615411.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013054.hg.1 |
ENST00000615411.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013054.hg.1 |
ENST00000615411.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000258106.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000394111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000464675.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000473732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000491023.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
ENST00000616281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008032.hg.1 |
NM_004097.2 |
Refseq |