Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
ENST00000526518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000328224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
ENST00000526518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010436.hg.1 |
NM_002233.3 |
Refseq |