Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304363.1 | NP_001291292.1 | 1 - 2 | 71 | 82 | |
| NM_020924.3 | NP_065975.1 | 1 - 2 | 86 | |||
| GenBank mRNA | BC018748.1 | - | 1 - 2 | 4553653 | 86 | |
| DA212731.1 | - | 1 - 2 | 71 | 82 | ||
| HY066289.1 | - | 1 - 2 | 86 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018748.1, NM_001304363.1, NM_020924.3 transcripts.
Set Membership: |
|

ZBTB26
Genomic Map