Assay Details
| Gene Symbol: ZNF629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080417.1 | NP_001073886.1 | 3 | 4468 | 62 | |
| XM_005255222.3 | XP_005255279.1 | 3 | 4607 | 62 | ||
| GenBank mRNA | AB002324.1 | - | 3 | 4434 | 62 | |
| BC080594.1 | - | 1 | 749 | 62 | ||
More Information
Set Membership: |
|

ZNF629
Genomic Map