Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024683.3 | NP_078959.3 | 4 | 892 | 82 | |
| XM_006722084.1 | XP_006722147.1 | 4 | 1376 | 82 | ||
| GenBank mRNA | AF086465.1 | - | 1 | 130 | 82 | |
| AJ314645.1 | - | 4 | 859 | 82 | ||
| AK026382.1 | - | 4 | 818 | 82 | ||
| AK225474.1 | - | 4 | 818 | 82 | ||
| BC009025.2 | - | 3 | 792 | 82 | ||
| BC073169.1 | - | 3 | 515 | 82 | ||
| BQ003763.1 | - | 1 | 82 | |||
| CR457360.1 | - | 2 | 216 | 82 | ||
More Information
Set Membership: |
|

TEFM
Genomic Map