Assay Details
| Gene Symbol: ANKRD27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032139.2 | NP_115515.2 | 29 | 3328 | 121 | |
| XM_006723412.2 | XP_006723475.1 | 29 | 3221 | 121 | ||
| XM_017027349.1 | XP_016882838.1 | 28 | 3190 | 121 | ||
| GenBank mRNA | AK054561.1 | - | 29 | 3317 | 121 | |
| AL136784.1 | - | 29 | 3305 | 121 | ||
| AL834335.1 | - | 10 | 1245 | 121 | ||
| BC045605.1 | - | 29 | 3333 | 121 | ||
| BC050529.1 | - | 29 | 3284 | 6067580 | 121 | |
More Information
Set Membership: |
|

ANKRD27
Genomic Map