Assay Details
| Gene Symbol: TENM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017017526.1 | XP_016873015.1 | 1 | 398 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017017526.1 transcripts.
Set Membership: |
|

TENM4
Genomic Map