Assay Details
| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011532697.2 | XP_011530999.1 | 1 | 2666 | 185 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011532697.2 transcripts.
Set Membership: |
|

EMX1
Genomic Map