Assay Details
| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007794.1 | NP_001007795.1 | 9 | 2089 | 103 | |
| NM_006090.3 | NP_006081.1 | 9 | 2051 | 103 | ||
| XM_006710285.3 | XP_006710348.1 | 9 | 2043 | 103 | ||
| GenBank mRNA | AK095229.1 | - | 1 | 2521 | 103 | |
| AL832104.1 | - | 4 | 5139 | 103 | ||
| BC032610.2 | - | 10 | 2051 | 5554219 | 103 | |
More Information
Set Membership: |
|

CEPT1
Genomic Map