Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534085.2 | XP_011532387.1 | 3 - 4 | 5508 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011534085.2 transcripts.
Set Membership: |
|

WNT5A
Genomic Map