Assay Details
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB037780.1 | - | 1 | 1762 | 86 | |
AB098731.1 | - | 1 | 375 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB098731.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
