Assay Details
| Gene Symbol: HHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263178.4 | XP_005263235.1 | 14 | 3693 | 71 | |
| XM_006714288.3 | XP_006714351.1 | 14 | 3689 | 71 | ||
More Information
Set Membership: |
|

HHIP
Genomic Map