Assay Details
| Gene Symbol: LOC105377792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017014118.1 | XP_016869607.1 | 4 - 5 | 791 | 127 | |
| XM_017014119.1 | XP_016869608.1 | 1 - 2 | 703 | 127 | ||
More Information
Set Membership: |
|

LOC105377792
Genomic Map