Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530923.2 | XP_011529225.1 | 7 | 2193 | 100 | |
| XM_017029450.1 | XP_016884939.1 | 7 | 1567 | 100 | ||
| XM_017029451.1 | XP_016884940.1 | 6 | 2064 | 100 | ||
More Information
Set Membership: |
|

YIPF6
Genomic Map