Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002359.3 | NP_002350.1 | 3 | 4573 | 122 | |
| NM_032711.3 | NP_116100.2 | 3 | 4605 | 122 | ||
| XM_011523578.1 | XP_011521880.1 | 3 | 5508 | 122 | ||
| GenBank mRNA | AK130699.1 | - | 1 | 1546 | 122 | |
| BC006143.1 | - | 1 | 956 | 122 | ||
| BC103722.1 | - | 1 | 1231 | 122 | ||
More Information
Set Membership: |
|

MAFG
Genomic Map