Assay Details
| Gene Symbol: C22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529907.2 | XP_011528209.1 | 1 | 353 | 98 | |
| XM_011529912.2 | XP_011528214.1 | 1 | 353 | 98 | ||
| XM_017028602.1 | XP_016884091.1 | 1 | 353 | 98 | ||
| XM_017028603.1 | XP_016884092.1 | 1 | 353 | 98 | ||
| XM_017028604.1 | XP_016884093.1 | 1 | 353 | 98 | ||
| XM_017028605.1 | XP_016884094.1 | 1 | 353 | 98 | ||
| XM_017028606.1 | XP_016884095.1 | 1 | 353 | 98 | ||
| XM_017028607.1 | XP_016884096.1 | 1 | 353 | 98 | ||
| XM_017028608.1 | XP_016884097.1 | 1 | 353 | 98 | ||
| XM_017028611.1 | XP_016884100.1 | 1 | 353 | 98 | ||
| XM_017028613.1 | XP_016884102.1 | 1 | 353 | 98 | ||
| XM_017028614.1 | XP_016884103.1 | 1 | 353 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529907.2, XM_011529912.2, XM_017028602.1, XM_017028603.1, XM_017028604.1, XM_017028605.1, XM_017028606.1, XM_017028607.1, XM_017028608.1, XM_017028611.1, XM_017028613.1, XM_017028614.1 transcripts.
Set Membership: |
|

C22orf15
Genomic Map