Assay Details
| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198560.2 | NP_940962.1 | 4 | 2982 | 81 | |
| XM_017006359.1 | XP_016861848.1 | 3 | 3520 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHFPL4| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198560.2 | NP_940962.1 | 4 | 2982 | 81 | |
| XM_017006359.1 | XP_016861848.1 | 3 | 3520 | 81 | ||
Set Membership: |
|