Assay Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 10439 | 92 | |
| NM_001164343.2 | NP_001157815.1 | 12 | 10901 | 92 | ||
| NM_001164344.2 | NP_001157816.1 | 8 | 10865 | 92 | ||
| NM_001164345.2 | NP_001157817.1 | 7 | 10819 | 92 | ||
| NM_001164346.2 | NP_001157818.1 | 6 | 10458 | 92 | ||
| NM_001164347.2 | NP_001157819.1 | 6 | 10480 | 92 | ||
| NM_015642.5 | NP_056457.3 | 10 | 10663 | 92 | ||
| NR_121662.1 | 7 | 8921 | 92 | |||
More Information
Set Membership: |
|

ZBTB20
Genomic Map