Assay Details
| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017008592.1 | XP_016864081.1 | 1 | 121 | 97 | |
| XM_017008599.1 | XP_016864088.1 | 1 | 121 | 97 | ||
| GenBank mRNA | DA031262.1 | - | 1 | 73 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017008592.1, XM_017008599.1 transcripts.
Set Membership: |
|

ARSJ
Genomic Map