Assay Details
| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014141.5 | NP_054860.1 | 24 | 8903 | 78 | |
| GenBank mRNA | CR933671.1 | - | 4 | 5083 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNTNAP2| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014141.5 | NP_054860.1 | 24 | 8903 | 78 | |
| GenBank mRNA | CR933671.1 | - | 4 | 5083 | 78 | |
Set Membership: |
|