Assay Details
| Gene Symbol: IPO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002271.4 | NP_002262.3 | 29 | 4951 | 76 | |
| XM_005254052.3 | XP_005254109.1 | 28 | 4861 | 76 | ||
| XM_005254053.3 | XP_005254110.1 | 28 | 4878 | 76 | ||
| XM_011521087.2 | XP_011519389.1 | 30 | 5042 | 76 | ||
| XM_011521088.2 | XP_011519390.1 | 30 | 5108 | 76 | ||
| XM_011521089.2 | XP_011519391.1 | 31 | 5165 | 76 | ||
| XM_011521090.2 | XP_011519392.1 | 29 | 5001 | 76 | ||
| XM_017020562.1 | XP_016876051.1 | 29 | 4982 | 76 | ||
| GenBank mRNA | AB074173.1 | - | 1 | 1083 | 76 | |
| AF339834.1 | - | 1 | 971 | 76 | ||
More Information
Set Membership: |
|

IPO5
Genomic Map