Assay Details
| Gene Symbol: RGS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102450.2 | NP_001095920.1 | 7 | 1571 | 86 | |
| NM_033345.3 | NP_203131.1 | 6 | 1442 | 86 | ||
| XM_005245555.3 | XP_005245612.1 | 7 | 1585 | 86 | ||
| XM_011510089.2 | XP_011508391.1 | 7 | 2482 | 86 | ||
| XM_011510090.2 | XP_011508392.1 | 10 | 2312 | 86 | ||
| XM_011510091.2 | XP_011508393.1 | 6 | 1459 | 86 | ||
| XM_017002631.1 | XP_016858120.1 | 10 | 1932 | 86 | ||
| XM_017002632.1 | XP_016858121.1 | 8 | 1763 | 86 | ||
| XM_017002633.1 | XP_016858122.1 | 7 | 1961 | 86 | ||
| XM_017002634.1 | XP_016858123.1 | 7 | 1640 | 86 | ||
| XM_017002635.1 | XP_016858124.1 | 8 | 2087 | 86 | ||
| XM_017002636.1 | XP_016858125.1 | 6 | 1569 | 86 | ||
| XM_017002637.1 | XP_016858126.1 | 7 | 1767 | 86 | ||
More Information
Set Membership: |
|

RGS8
Genomic Map