Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024591.4 | NP_078867.2 | 8 | 1543 | 62 | |
| GenBank mRNA | AK021811.1 | - | 8 | 1481 | 62 | |
| AK223187.1 | - | 8 | 1505 | 62 | ||
| BC010108.2 | - | 8 | 1489 | 3604039 | 62 | |
More Information
Set Membership: |
|

CHMP6
Genomic Map