Assay Details
| Gene Symbol: GACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126369.1 | 1 - 2 | 156 | 78 | ||
| NR_126370.1 | 1 - 2 | 156 | 78 | |||
| GenBank mRNA | DB077273.1 | - | 1 - 2 | 123 | 78 | |
| DB096621.1 | - | 1 - 2 | 156 | 78 | ||
More Information
Set Membership: |
|

GACAT1
Genomic Map