Assay Details
| Gene Symbol: SMIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317896.1 | NP_001304825.1 | 6 | 3139 | 80 | |
| NM_001317897.1 | NP_001304826.1 | 5 | 2848 | 80 | ||
| NM_174921.2 | NP_777581.1 | 5 | 2994 | 80 | ||
| XM_011513640.2 | XP_011511942.1 | 5 | 3127 | 80 | ||
| XM_017007862.1 | XP_016863351.1 | 6 | 3043 | 80 | ||
| XM_017007863.1 | XP_016863352.1 | 5 | 2884 | 80 | ||
More Information
Set Membership: |
|

SMIM14
Genomic Map