Assay Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006425.4 | NP_006416.3 | 2 | 264 | 116 | |
| XM_005265793.3 | XP_005265850.1 | 2 | 611 | 116 | ||
| XM_011534418.2 | XP_011532720.1 | 3 | 314 | 116 | ||
| GenBank mRNA | AF101074.1 | - | 1 | 109 | 116 | |
| AK024875.1 | - | 2 | 184 | 116 | ||
| AK314564.1 | - | 2 | 184 | 116 | ||
| AY486334.1 | - | 2 | 144 | 116 | ||
| BC010634.1 | - | 2 | 190 | 3871882 | 116 | |
| DA409854.1 | - | 2 | 190 | 116 | ||
| DQ174516.1 | - | 2 | 196 | 116 | ||
| DQ174517.1 | - | 2 | 234 | 116 | ||
More Information
Set Membership: |
|

SLU7
Genomic Map