Assay Details
| Gene Symbol: CDK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002596.3 | NP_002587.2 | 1 | 138 | 65 | |
| NM_212502.2 | NP_997667.1 | 1 | 138 | 65 | ||
| NM_212503.2 | NP_997668.1 | 1 | 138 | 65 | ||
| GenBank mRNA | AK293756.1 | - | 1 | 97 | 65 | |
| AK295654.1 | - | 1 | 98 | 65 | ||
| AK296320.1 | - | 1 | 96 | 65 | ||
| AU130437.1 | - | 1 | 99 | 65 | ||
| AY353237.1 | - | 1 | 53 | 65 | ||
| AY353240.1 | - | 1 | 35 | 65 | ||
| BM772779.1 | - | 1 | 95 | 65 | ||
| BU165355.1 | - | 58 | 65 | |||
| DC381358.1 | - | 1 | 141 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293756.1, AK295654.1, AK296320.1, AY353237.1, NM_002596.3, NM_212502.2, NM_212503.2 transcripts.
Set Membership: |
|

CDK18
Genomic Map