Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016052.3 | NP_057136.2 | 1 | 66 | 81 | |
| XM_011509597.2 | XP_011507899.1 | 1 | 83 | 81 | ||
| GenBank mRNA | AK290066.1 | - | 1 | 48 | 81 | |
| BC003668.1 | - | 1 | 44 | 81 | ||
| BC020641.1 | - | 1 | 43 | 4772128 | 81 | |
| BF242786.1 | - | 1 | 67 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK290066.1, BC020641.1 transcripts.
Set Membership: |
|

RRP15
Genomic Map