Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199922.1 | NP_001186851.1 | 12 | 2267 | 68 | |
| NM_170601.4 | NP_733746.1 | 10 | 2136 | 68 | ||
| XM_017017930.1 | XP_016873419.1 | 11 | 4656 | 68 | ||
| GenBank mRNA | AF303378.1 | - | 10 | 1975 | 68 | |
| AL137496.1 | - | 10 | 1958 | 68 | ||
| BC022456.1 | - | 10 | 1990 | 68 | ||
| BC068450.1 | - | 10 | 2122 | 5278370 | 68 | |
More Information
Set Membership: |
|

SIAE
Genomic Map