Assay Details
| Gene Symbol: ACOT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152331.3 | NP_689544.3 | 1 | 289 | 88 | |
| GenBank mRNA | AK094223.1 | - | 1 | 58 | 88 | |
| BC090945.1 | - | 1 | 289 | 5189326 | 88 | |
| BC117341.1 | - | 1 | 68 | 40125892 | 88 | |
| BC117343.1 | - | 1 | 68 | 40125894 | 88 | |
| BX248023.1 | - | 1 | 124 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK094223.1, BC090945.1, BC117341.1, BC117343.1, BX248023.1, NM_152331.3 transcripts.
Set Membership: |
|

ACOT4
Genomic Map