Assay Details
| Gene Symbol: LRFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024036.4 | NP_076941.2 | 1 | 1451 | 64 | |
| XM_005274239.3 | XP_005274296.1 | 2 | 1331 | 64 | ||
| GenBank mRNA | AB209005.1 | - | 1 | 1072 | 64 | |
| BC000207.1 | - | 1 | 188 | 64 | ||
| BC007718.2 | - | 1 | 1331 | 64 | ||
| BC014040.2 | - | 1 | 195 | 64 | ||
| BC015581.2 | - | 1 | 1354 | 4637053 | 64 | |
| BC027475.1 | - | 1 | 790 | 64 | ||
| BC071866.1 | - | 2 | 1300 | 64 | ||
| BC094813.1 | - | 1 | 529 | 64 | ||
More Information
Set Membership: |
|

LRFN4
Genomic Map