Assay Details
| Gene Symbol: SORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003105.5 | NP_003096.1 | 26 | 3781 | 100 | |
| XM_011542963.2 | XP_011541265.1 | 25 | 3542 | 100 | ||
| XM_011542965.2 | XP_011541267.1 | 20 | 2690 | 100 | ||
| XM_011542967.2 | XP_011541269.1 | 6 | 722 | 100 | ||
| XM_017018169.1 | XP_016873658.1 | 26 | 3488 | 100 | ||
| XM_017018170.1 | XP_016873659.1 | 24 | 3164 | 100 | ||
| XM_017018171.1 | XP_016873660.1 | 26 | 3656 | 100 | ||
| XM_017018172.1 | XP_016873661.1 | 12 | 1653 | 100 | ||
| GenBank mRNA | AK295090.1 | - | 3 | 571 | 100 | |
| AK295878.1 | - | 6 | 722 | 100 | ||
| AK302589.1 | - | 3 | 393 | 100 | ||
| BC137171.1 | - | 26 | 3704 | 9021168 | 100 | |
| Y08110.1 | - | 26 | 3732 | 100 | ||
More Information
Set Membership: |
|

SORL1
Genomic Map