Assay Details
| Gene Symbol: TIPARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184717.1 | NP_001171646.1 | 2 | 815 | 90 | |
| NM_001184718.1 | NP_001171647.1 | 2 | 380 | 90 | ||
| NM_015508.4 | NP_056323.2 | 2 | 730 | 90 | ||
| GenBank mRNA | AK097785.1 | - | 2 | 559 | 90 | |
| AK291261.1 | - | 2 | 565 | 90 | ||
| BC034397.1 | - | 2 | 380 | 90 | ||
| BC050350.1 | - | 2 | 537 | 5264807 | 90 | |
| BX537965.1 | - | 2 | 557 | 90 | ||
| CR749647.1 | - | 2 | 816 | 90 | ||
More Information
Set Membership: |
|

TIPARP
Genomic Map