Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135179.1 | NP_001128651.1 | 7 | 8846 | 91 | |
| NM_016598.2 | NP_057682.1 | 8 | 8930 | 91 | ||
| XM_017006563.1 | XP_016862052.1 | 9 | 1289 | 91 | ||
| GenBank mRNA | AK127837.1 | - | 1 | 404 | 91 | |
More Information
Set Membership: |
|

ZDHHC3
Genomic Map