Assay Details
| Gene Symbol: ZFYVE16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284237.1 | NP_001271166.1 | 1 - 2 | 136 | 87 | |
| NM_014733.4 | NP_055548.3 | 1 - 2 | 136 | 87 | ||
| GenBank mRNA | BC030808.1 | - | 1 - 2 | 66 | 4701661 | 87 |
| BX537424.1 | - | 1 - 2 | 88 | 87 | ||
| DA216169.1 | - | 1 - 2 | 136 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030808.1, BX537424.1, NM_001284237.1, NM_014733.4 transcripts.
Set Membership: |
|

ZFYVE16
Genomic Map