Assay Details
| Gene Symbol: ZFYVE16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105251.2 | NP_001098721.1 | 3 - 4 | 379 | 98 | |
| NM_001284236.1 | NP_001271165.1 | 3 - 4 | 305 | 98 | ||
| NM_001284237.1 | NP_001271166.1 | 2 - 3 | 251 | 98 | ||
| NM_014733.4 | NP_055548.3 | 2 - 3 | 251 | 98 | ||
| XM_005248632.4 | XP_005248689.1 | 2 - 3 | 377 | 98 | ||
| XM_011543753.2 | XP_011542055.1 | 3 - 4 | 276 | 98 | ||
| XM_017010091.1 | XP_016865580.1 | 4 - 5 | 437 | 98 | ||
| XM_017010092.1 | XP_016865581.1 | 3 - 4 | 423 | 98 | ||
| XM_017010095.1 | XP_016865584.1 | 3 - 4 | 276 | 98 | ||
| GenBank mRNA | AB002303.1 | - | 3 - 4 | 319 | 98 | |
| AF434817.1 | - | 1 - 2 | 71 | 98 | ||
| BC030808.1 | - | 2 - 3 | 181 | 4701661 | 98 | |
| BX537424.1 | - | 2 - 3 | 203 | 98 | ||
| BX956000.1 | - | 2 - 3 | 250 | 98 | ||
| CR933621.1 | - | 3 - 4 | 254 | 98 | ||
| DA216169.1 | - | 2 - 3 | 251 | 98 | ||
| DR005686.1 | - | 3 - 4 | 246 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010095.1 transcripts.
Set Membership: |
|

ZFYVE16
Genomic Map