Assay Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286550.1 | NP_001273479.1 | 2 - 3 | 235 | 123 | |
| GenBank mRNA | AY358823.1 | - | 2 - 3 | 214 | 123 | |
More Information
Set Membership: |
|

CLPSL2
Genomic Map