Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042762.2 | NP_001036227.1 | 3 - 4 | 361 | 101 | |
| NM_001287492.1 | NP_001274421.1 | 3 - 4 | 799 | 101 | ||
| NM_001287493.1 | NP_001274422.1 | 2 - 3 | 193 | 101 | ||
| NM_001287494.1 | NP_001274423.1 | 2 - 3 | 188 | 101 | ||
| NM_001287496.1 | NP_001274425.1 | 2 - 3 | 193 | 101 | ||
| NM_022116.4 | NP_071399.2 | 3 - 4 | 354 | 101 | ||
| XM_011515470.2 | XP_011513772.1 | 3 - 4 | 363 | 101 | ||
| GenBank mRNA | AK023142.1 | - | 3 - 4 | 314 | 101 | |
| AK027537.1 | - | 3 - 4 | 761 | 101 | ||
| BC051867.1 | - | 3 - 4 | 295 | 6158883 | 101 | |
| DA114757.1 | - | 2 - 3 | 191 | 101 | ||
| DA121992.1 | - | 2 - 3 | 145 | 101 | ||
| DA433617.1 | - | 2 - 3 | 155 | 101 | ||
| DA812999.1 | - | 2 - 3 | 273 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK023142.1, AK027537.1, BC051867.1, NM_001042762.2, NM_001287492.1, NM_001287493.1, NM_001287494.1, NM_001287496.1, NM_022116.4, XM_011515470.2 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map