Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287495.1 | NP_001274424.1 | 1 - 2 | 89 | 76 | |
| GenBank mRNA | DA121143.1 | - | 1 - 2 | 46 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287495.1 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map