Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002901.3 | NP_001002901.1 | 3 - 4 | 261 | 75 | |
| NM_001288829.1 | NP_001275758.1 | 1 - 2 | 47 | 75 | ||
| NM_001288831.1 | NP_001275760.1 | 1 - 2 | 47 | 75 | ||
| NM_001320241.1 | NP_001307170.1 | 2 - 3 | 75 | 75 | ||
| GenBank mRNA | AK290733.1 | - | 2 - 3 | 75 | 75 | |
| AY670683.1 | - | 1 - 2 | 42 | 75 | ||
| AY670684.1 | - | 1 - 2 | 42 | 75 | ||
| AY670686.1 | - | 1 - 2 | 42 | 75 | ||
| DQ021957.1 | - | 3 - 4 | 261 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK290733.1, AY670683.1, AY670684.1, AY670686.1, DQ021957.1, NM_001002901.3, NM_001288829.1, NM_001288831.1, NM_001320241.1 transcripts.
Set Membership: |
|

FCRLB
Genomic Map