Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002901.3 | NP_001002901.1 | 4 - 5 | 287 | 99 | |
| NM_001288829.1 | NP_001275758.1 | 2 - 3 | 73 | 99 | ||
| NM_001288831.1 | NP_001275760.1 | 2 - 3 | 73 | 99 | ||
| NM_001320241.1 | NP_001307170.1 | 3 - 4 | 101 | 99 | ||
| GenBank mRNA | AK290733.1 | - | 3 - 4 | 101 | 99 | |
| AY670683.1 | - | 2 - 3 | 68 | 99 | ||
| AY670684.1 | - | 2 - 3 | 68 | 99 | ||
| AY670686.1 | - | 2 - 3 | 68 | 99 | ||
| AY786314.1 | - | 2 - 3 | 53 | 99 | ||
| DQ021957.1 | - | 4 - 5 | 287 | 99 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map