Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288829.1 | NP_001275758.1 | 4 - 5 | 585 | 68 | |
| NM_001288830.1 | NP_001275759.1 | 3 - 4 | 564 | 68 | ||
| GenBank mRNA | AY670684.1 | - | 4 - 5 | 580 | 68 | |
| AY670685.1 | - | 3 - 4 | 559 | 68 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map