Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288830.1 | NP_001275759.1 | 1 - 2 | 51 | 100 | |
| NM_001288832.1 | NP_001275761.1 | 1 - 2 | 51 | 100 | ||
| GenBank mRNA | BX364259.2 | - | 3 - 4 | 246 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001288830.1, NM_001288832.1 transcripts.
Set Membership: |
|

FCRLB
Genomic Map