Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288831.1 | NP_001275760.1 | 4 - 5 | 594 | 79 | |
| NM_001288832.1 | NP_001275761.1 | 3 - 4 | 573 | 79 | ||
| GenBank mRNA | AY670686.1 | - | 4 - 5 | 589 | 79 | |
| AY670687.1 | - | 3 - 4 | 568 | 79 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map