Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304364.1 | NP_001291293.1 | 1 - 2 | 208 | 77 | |
| GenBank mRNA | AK057445.1 | - | 1 - 2 | 208 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK057445.1, NM_001304364.1 transcripts.
Set Membership: |
|

ZBTB26
Genomic Map