Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013625.3 | NP_001013647.2 | 2 - 3 | 96 | 95 | |
| GenBank mRNA | DT215043.1 | - | 2 - 3 | 96 | 95 | |
More Information
Set Membership: |
|

CFAP126
Genomic Map