Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013625.3 | NP_001013647.2 | 4 - 5 | 362 | 97 | |
| GenBank mRNA | BU688150.1 | - | 1 - 2 | 439 | 97 | |
| DT215043.1 | - | 4 - 5 | 362 | 97 | ||
More Information
Set Membership: |
|

CFAP126
Genomic Map