Assay Details
| Gene Symbol: C6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 314 | 81 | |
| GenBank mRNA | AF451991.1 | - | 1 | 265 | 81 | |
| AK295061.1 | - | 1 | 300 | 81 | ||
| BC051700.1 | - | 1 | 265 | 4792634 | 81 | |
| CF994905.1 | - | 341 | 81 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF451991.1, AK295061.1, BC051700.1, NM_001029863.2 transcripts.
Set Membership: |
|

C6orf120
Genomic Map