Assay Details
| Gene Symbol: FAM24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029888.1 | NP_001025059.1 | 1 - 2 | 119 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001029888.1 transcripts.
Set Membership: |
|

FAM24A
Genomic Map