Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320241.1 | NP_001307170.1 | 1 - 2 | 25 | 56 | |
| GenBank mRNA | AK290733.1 | - | 1 - 2 | 25 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK290733.1, NM_001320241.1 transcripts.
Set Membership: |
|

FCRLB
Genomic Map